accessibility menu, dialog, popup

UserName

Viewing profile as user:
FNI

CAS RN 22282-75-3

IUPAC Name: 3-fluoro-4-iodopyridine
Synonyms: 3-fluoro-4-iodopyridine
Molecular Weight (g/mol): 222.99
Molecular Formula: C5H3FIN
InChi Key: MSLUTDVOKZOXTA-UHFFFAOYSA-N
SMILES: FC1=CN=CC=C1I
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)