accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HCl

CAS RN 22287-35-0

IUPAC Name: bicyclo[1.1.1]pentan-1-amine hydrochloride
Synonyms: bicyclo[1.1.1]pentan-1-amine hydrochloride
Molecular Weight (g/mol): 119.59
Molecular Formula: C5H10ClN
InChi Key: LQKLVOWNBKJRJE-UHFFFAOYSA-N
SMILES: Cl.NC12CC(C1)C2
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)