accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOH3CCH3OSOOCH3

CAS RN 22308-12-9

IUPAC Name: 1-({2,2-dimethyl-3-[(4-methylbenzenesulfonyl)oxy]propoxy}sulfonyl)-4-methylbenzene
Synonyms: 1-{2,2-dimethyl-3-[(4-methylbenzenesulfonyl)oxy]propoxysulfonyl}-4-methylbenzene
Molecular Weight (g/mol): 412.52
Molecular Formula: C19H24O6S2
InChi Key: LJISGCCLDDOYIJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC(C)(C)COS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results