accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClClClCl

CAS RN 2234-13-1

IUPAC Name: octachloronaphthalene
Synonyms: perna
Molecular Weight (g/mol): 403.71
Molecular Formula: C10Cl8
InChi Key: RTNLUFLDZOAXIC-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C2=C(Cl)C(Cl)=C(Cl)C(Cl)=C2C(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results