accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONO

CAS RN 22346-75-4

IUPAC Name: 4-(hydroxymethyl)pyridin-1-ium-1-olate
Synonyms: 4-(hydroxymethyl)pyridin-1-ium-1-olate
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: ZJGXPKKVSNSGRJ-UHFFFAOYSA-N
SMILES: OCC1=CC=[N+]([O-])C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results