accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3COHNOHHNOO

CAS RN 223472-31-9

IUPAC Name: 5-(ethoxymethoxy)-N-hydroxy-2-methyl-4-[(4-phenoxyphenyl)formamido]pentanamide
Synonyms: 5-(ethoxymethoxy)-N-hydroxy-2-methyl-4-[(4-phenoxyphenyl)formamido]pentanamide
Molecular Weight (g/mol): 416.47
Molecular Formula: C22H28N2O6
InChi Key: HDWWQELUBWGQGA-UHFFFAOYNA-N
SMILES: CCOCOCC(CC(C)C(=O)NO)NC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results