accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2235-54-3

NH4H3COSOOO

CAS RN 2235-54-3

IUPAC Name: ammonium dodecyl sulfate
Synonyms: ammonium lauryl sulfate
Molecular Weight (g/mol): 283.43
Molecular Formula: C12H29NO4S
InChi Key: BTBJBAZGXNKLQC-UHFFFAOYSA-N
SMILES: [NH4+].CCCCCCCCCCCCOS([O-])(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results