accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONHOCl

CAS RN 22353-38-4

IUPAC Name: 3-chloro-5-nitro-1,2-dihydropyridin-2-one
Synonyms: 3-chloro-5-nitro-1H-pyridin-2-one
Molecular Weight (g/mol): 174.54
Molecular Formula: C5H3ClN2O3
InChi Key: GTOXJMNILNYXLE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CNC(=O)C(Cl)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results