Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-5-nitro-2-pyridone 98.0+%, TCI America™
SDP

Catalog No. C31851G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C31851G Supplier TCI America Supplier No. C31851G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22353-38-4
Molecular Formula C5H3ClN2O3
Molecular Weight (g/mol) 174.54
MDL Number MFCD17015099
InChI Key GTOXJMNILNYXLE-UHFFFAOYSA-N
Synonym 3-Chloro-2-hydroxy-5-nitropyridine, 3-Chloro-5-nitro-2-pyridinol
PubChem CID 336289
IUPAC Name 3-chloro-5-nitro-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1[N+](=O)[O-])Cl

Specifications

Melting Point 197°C
Color Yellow
Quantity 1 g
Formula Weight 174.54
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-5-nitro-2-pyridone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.