accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2237-36-7

IUPAC Name: 2-hydroxy-4-methoxybenzoic acid
Synonyms: 2-hydroxy-4-methoxybenzoic acid
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: MRIXVKKOHPQOFK-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(O)=O)C(O)=C1
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)