accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22381-54-0

IUPAC Name: 2-(4-methylbenzenesulfonyl)ethan-1-ol
Synonyms: 2-(4-methylbenzenesulfonyl)ethanol
Molecular Weight (g/mol): 200.25
Molecular Formula: C9H12O3S
InChi Key: QJFIXBNLKARINT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)CCO
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results