Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(p-Toluenesulfonyl)ethanol 95.0+%, TCI America™
SDP

Catalog No. T16205G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16205G Supplier TCI America Supplier No. T16205G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22381-54-0
Molecular Formula C9H12O3S
Molecular Weight (g/mol) 200.252
MDL Number MFCD00041757
InChI Key QJFIXBNLKARINT-UHFFFAOYSA-N
Synonym 2-tosylethanol, 2-p-toluenesulfonyl ethanol, 2-p-tolylsulfonyl ethanol, 2-4-methylphenyl sulfonyl ethanol, ethanol, 2-p-tolylsulfonyl, ethanol, 2-4-methylphenyl sulfonyl, 2-4-methylphenylsulfonyl ethanol, 2-hydroxyethyl 4-methylphenyl sulfone, 2-4-methylbenzenesulfonyl ethanol
PubChem CID 89682
IUPAC Name 2-(4-methylphenyl)sulfonylethanol
SMILES CC1=CC=C(C=C1)S(=O)(=O)CCO

Specifications

Melting Point 55°C
Boiling Point 195°C
Quantity 5 g
Formula Weight 200.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(p-Toluenesulfonyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.