accessibility menu, dialog, popup

UserName

Viewing profile as user:
HoBrBrBr

CAS RN 223911-98-6

IUPAC Name: holmium(3+) tribromide
Synonyms: holmium(III) bromide
Molecular Weight (g/mol): 404.64
Molecular Formula: Br3Ho
InChi Key: MZNSYJWLQLXLHE-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[Ho+3]
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results