accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22426-47-7

IUPAC Name: 6-(acetyloxy)naphthalen-2-yl acetate
Synonyms: 6-(acetyloxy)naphthalen-2-yl acetate
Molecular Weight (g/mol): 244.25
Molecular Formula: C14H12O4
InChi Key: JTUIDPCUTRXCPW-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC2=CC=C(OC(C)=O)C=C2C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results