Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diacetoxynaphthalene 99.0+%, TCI America™
SDP

Catalog No. D17395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D17395G Supplier TCI America Supplier No. D17395G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22426-47-7
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD00035955
InChI Key JTUIDPCUTRXCPW-UHFFFAOYSA-N
PubChem CID 594930
IUPAC Name (6-acetyloxynaphthalen-2-yl) acetate
SMILES CC(=O)OC1=CC2=C(C=C1)C=C(C=C2)OC(=O)C

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 5 g
Formula Weight 244.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diacetoxynaphthalene
EINECSNumber (4)-1602
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.