CAS RN 2242743-87-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Lenalidomide 5'-piperazine-7'-methoxy
Cereblon ligand with terminal piperazine for onward chemistry