accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2243-83-6

IUPAC Name: naphthalene-2-carbonyl chloride
Synonyms: 2-naphthalenecarbonyl chloride
Molecular Weight (g/mol): 190.63
Molecular Formula: C11H7ClO
InChi Key: XNLBCXGRQWUJLU-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (6)
Boiling Point 
  • (3)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (1)
  • (5)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)