Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

βNaphthoyl Chloride, MP Biomedicals

Catalog No. ICN216251 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN216251 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN216251 Supplier MP Biomedicals, Inc Supplier No. 0521625110

Chemical Identifiers

CAS 2243-83-6
Molecular Formula C11H7ClO
Molecular Weight (g/mol) 190.626
InChI Key XNLBCXGRQWUJLU-UHFFFAOYSA-N
Synonym 2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride
PubChem CID 75246
IUPAC Name naphthalene-2-carbonyl chloride
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)Cl

Specifications

Quantity 10 g
Formula Weight 190.63
Chemical Name or Material Beta-Naphthoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.