accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2244386-63-6

IUPAC Name: sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
Synonyms: pravastatin sodium
Molecular Weight (g/mol): 446.52
Molecular Formula: C23H35NaO7
InChi Key: VWBQYTRBTXKKOG-IYNICTALSA-M
SMILES: [Na+].[H][C@]12[C@H](C[C@H](O)C=C1C=C[C@H](C)[C@@H]2CC[C@@H](O)C[C@@H](O)CC([O-])=O)OC(=O)[C@@H](C)CC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)