Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Pravastatin sodium salt hydrate

Catalog No. AC464882500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464882500 250 mg
AC464880010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC464882500 Supplier Thermo Scientific Chemicals Supplier No. 464882500
  • $178.80
    $151.65 / Each of 1

    Save $27.15

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2244386-63-6 | C23H35NaO7 | 446.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2244386-63-6
Molecular Formula C23H35NaO7
Molecular Weight (g/mol) 446.52
MDL Number MFCD00887601
InChI Key VWBQYTRBTXKKOG-IYNICTALSA-M
IUPAC Name sodium (3R,5R)-7-[(1S,2S,6S,8S,8aR)-6-hydroxy-2-methyl-8-{[(2S)-2-methylbutanoyl]oxy}-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate
SMILES [Na+].[H][C@]12[C@H](C[C@H](O)C=C1C=C[C@H](C)[C@@H]2CC[C@@H](O)C[C@@H](O)CC([O-])=O)OC(=O)[C@@H](C)CC

Specifications

Melting Point 171.0°C to 173.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 446.51
Chemical Name or Material Pravastatin sodium salt hydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Suspected of damaging fertility or the unborn child.

May cause harm to breast-fed children.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

Avoid contact during pregnancy and while nursing

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.