accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2245-30-9

IUPAC Name: 2-tert-butyloxirane
Synonyms: 2-tert-butyloxirane
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: HEAYDCIZOFDHRM-UHFFFAOYNA-N
SMILES: CC(C)(C)C1CO1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)