accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2245-52-5

IUPAC Name: [(4-ethenylphenyl)methyl]dimethylazanium
Synonyms: [(4-ethenylphenyl)methyl]dimethylazanium
Molecular Weight (g/mol): 162.26
Molecular Formula: C11H16N
InChi Key: XQBHAZDVLGNSOJ-UHFFFAOYSA-O
SMILES: C[NH+](C)CC1=CC=C(C=C)C=C1
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results