accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 224779-27-5

IUPAC Name: tert-butyl 3-hydroxy-1,2,3,6-tetrahydropyridine-1-carboxylate
Synonyms: tert-butyl 3-hydroxy-3,6-dihydro-2H-pyridine-1-carboxylate
Molecular Weight (g/mol): 199.25
Molecular Formula: C10H17NO3
InChi Key: NICJOYHYEDFTIX-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC=CC(O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)