accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOHOFF

CAS RN 22494-42-4

IUPAC Name: 2',4'-difluoro-4-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
Synonyms: diflunisal difluorophenylsalicylic acid dolisal frosst SA brand of diflunisal noaldol reuflos diflunisal novopharm brand diflunisalum flustar nu-diflunisal
Molecular Weight (g/mol): 250.2
Molecular Formula: C13H8F2O3
InChi Key: HUPFGZXOMWLGNK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1O)C1=CC=C(F)C=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results