accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 225656-59-7

IUPAC Name: 1-(chloromethyl)-2-nitro-4-(trifluoromethyl)benzene
Synonyms: 1-(chloromethyl)-2-nitro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 239.58
Molecular Formula: C8H5ClF3NO2
InChi Key: JWFRBTUSUAFGOS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(CCl)C=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results