accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHN

CAS RN 22580-55-8

IUPAC Name: pyrrolidin-2-one
Synonyms: pyrrolidone 2-pyrol aminobutyric lactam 2-pyrrolidone-butyrolactam gamma-aminobutyric lactam 2-pyrrolidone, hydrotribromide gamma-aminobutyric acid lactam M-pyrol hydrobromide 2-pyrrolidone LAM
Molecular Weight (g/mol): 85.11
Molecular Formula: C4H7NO
InChi Key: HNJBEVLQSNELDL-UHFFFAOYSA-N
SMILES: O=C1CCCN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results