accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3Cl

CAS RN 22592-15-0

IUPAC Name: 1-chloropent-2-yne
Synonyms: 1-chloropent-2-yne
Molecular Weight (g/mol): 102.56
Molecular Formula: C5H7Cl
InChi Key: JBKXDMAPOZZDCE-UHFFFAOYSA-N
SMILES: CCC#CCCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results