Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pentynyl Chloride 96.0+%, TCI America™
SDP

Catalog No. P11415ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11415ML 5 mL
P114125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11415ML Supplier TCI America Supplier No. P11415ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22592-15-0
Molecular Formula C5H7Cl
Molecular Weight (g/mol) 102.561
MDL Number MFCD00191678
InChI Key JBKXDMAPOZZDCE-UHFFFAOYSA-N
Synonym 1-Chloro-2-pentyne
PubChem CID 89764
IUPAC Name 1-chloropent-2-yne
SMILES CCC#CCCl

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 mL
UN Number 1993
Formula Weight 102.56
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pentynyl Chloride
EINECSNumber (2)-3854
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.