accessibility menu, dialog, popup

UserName

Viewing profile as user:
N

CAS RN 226-36-8

IUPAC Name: 2-azapentacyclo[12.8.0.0³,¹².0⁴,⁹.0¹⁵,²⁰]docosa-1,3(12),4,6,8,10,13,15,17,19,21-undecaene
Synonyms: dibenz(A,H)acridine
Molecular Weight (g/mol): 279.34
Molecular Formula: C21H13N
InChi Key: JNCSIWAONQTVCF-UHFFFAOYSA-N
SMILES: C1=CC=C2C(C=CC3=NC4=C(C=CC5=CC=CC=C45)C=C23)=C1
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results