accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOO

CAS RN 22651-87-2

IUPAC Name: cyclohexanecarbonyl cyclohexanecarboxylate
Synonyms: cyclohexanecarbonyl cyclohexanecarboxylate
Molecular Weight (g/mol): 238.33
Molecular Formula: C14H22O3
InChi Key: JOHUAELJNSBTGS-UHFFFAOYSA-N
SMILES: O=C(OC(=O)C1CCCCC1)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results