Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclohexanecarboxylic Anhydride 98.0+%, TCI America™
SDP

Catalog No. C27355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27355G Supplier TCI America Supplier No. C27355G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22651-87-2
Molecular Formula C14H22O3
Molecular Weight (g/mol) 238.33
MDL Number MFCD12404944
InChI Key JOHUAELJNSBTGS-UHFFFAOYSA-N
PubChem CID 89781
IUPAC Name cyclohexanecarbonyl cyclohexanecarboxylate
SMILES O=C(OC(=O)C1CCCCC1)C1CCCCC1

Specifications

Color Colorless
Boiling Point 156°C
Quantity 5 g
Formula Weight 238.33
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Cyclohexanecarboxylic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.