accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22658-92-0

IUPAC Name: octan-3-ol
Synonyms: 3-octanol octanol (isomer mix)
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: NMRPBPVERJPACX-UHFFFAOYNA-N
SMILES: CCCCCC(O)CC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)