Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-3-Octanol, 96%, Thermo Scientific™

Catalog No. AAH6696314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6696314 25 g
AAH6696306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6696314 Supplier Thermo Scientific Chemicals Supplier No. H6696314

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22658-92-0
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.231
MDL Number MFCD06797294
InChI Key NMRPBPVERJPACX-QMMMGPOBSA-N
Synonym s-+-3-octanol, 3s-octan-3-ol, octan-3s-ol, s-octan-3-ol, 3s-3-octanol, 3-octanol, 3s, unii-73dz0u3u1e component
PubChem CID 6999002
IUPAC Name (3S)-octan-3-ol
SMILES CCCCCC(CC)O

Specifications

Color Colorless to Yellow
Density 0.819 g/mL
Boiling Point 174°C to 177°C
Flash Point 68°C (154°F)
Refractive Index 1.425
Quantity 25 g
Formula Weight 130.23
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material (S)-(+)-3-Octanol
Hazard Category H227-H315-H319-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.