accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOSiCH3CH3H3CSiCH3CH3H3C

CAS RN 22737-33-3

IUPAC Name: 2,2,4,5,5-pentamethyl-3-oxa-4-aza-2,5-disilahexane
Synonyms: 2,2,4,5,5-pentamethyl-3-oxa-4-aza-2,5-disilahexane
Molecular Weight (g/mol): 191.42
Molecular Formula: C7H21NOSi2
InChi Key: LXNSJZVQLMUDMT-UHFFFAOYSA-N
SMILES: CN(O[Si](C)(C)C)[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results