Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N,O-bis(trimethylsilyl)hydroxylamine, TCI America™
SDP

Catalog No. M10771ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10771ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M10771ML Supplier TCI America Supplier No. M10771ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22737-33-3
Molecular Formula C7H21NOSi2
Molecular Weight (g/mol) 191.42
MDL Number MFCD00191463
InChI Key LXNSJZVQLMUDMT-UHFFFAOYSA-N
PubChem CID 12635082
IUPAC Name 2,2,4,5,5-pentamethyl-3-oxa-4-aza-2,5-disilahexane
SMILES CN(O[Si](C)(C)C)[Si](C)(C)C

Specifications

Color Yellow
Quantity 1 mL
UN Number 2920
Formula Weight 191.42
Physical Form Liquid
Chemical Name or Material N-Methyl-N,O-bis(trimethylsilyl)hydroxylamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.