CAS RN 2274-58-0
CAS RN 2274-58-0
IUPAC Name:
2,5-di{[(benzyloxy)carbonyl]amino}pentanoic acid
Synonyms:
2,5-di{[(benzyloxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol):
400.43
Molecular Formula:
C21H24N2O6
InChi Key:
VBENHRFIEOLOJJ-UHFFFAOYNA-N
SMILES:
OC(=O)C(CCCNC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
Advanced Chem Tech Z-Orn Z , Advanced ChemTech
CAS: 2274-58-0 Molecular Formula: C21H24N2O6 Molecular Weight (g/mol): 400.43 MDL Number: MFCD00037821,MFCD00339525 InChI Key: VBENHRFIEOLOJJ-UHFFFAOYNA-N IUPAC Name: 2,5-di{[(benzyloxy)carbonyl]amino}pentanoic acid SMILES: OC(=O)C(CCCNC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1