accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaCH3SOOO

CAS RN 22767-49-3

IUPAC Name: sodium pentane-1-sulfonate
Synonyms: sodium pentane-1-sulfonate
Molecular Weight (g/mol): 174.19
Molecular Formula: C5H11NaO3S
InChi Key: ROBLTDOHDSGGDT-UHFFFAOYSA-M
SMILES: [Na+].CCCCCS([O-])(=O)=O
Quantity 
  • (1)
  • (7)
  • (7)
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (9)
  • (3)
  • (1)
  • (1)
Grade 
  • (3)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (6)
  • (9)
  • (1)
  • (1)
  • (7)
  • (7)
  • (3)
  • (2)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (9)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)