accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOOOH3CH3CO

CAS RN 22781-23-3

IUPAC Name: 2,2-dimethyl-2H-1,3-benzodioxol-4-yl N-methylcarbamate
Synonyms: bendiocarb
Molecular Weight (g/mol): 223.23
Molecular Formula: C11H13NO4
InChi Key: XEGGRYVFLWGFHI-UHFFFAOYSA-N
SMILES: CNC(=O)OC1=CC=CC2=C1OC(C)(C)O2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results