accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHN(R)NHOHO

CAS RN 2280-01-5

IUPAC Name: (2R)-2-acetamido-3-(1H-indol-3-yl)propanoic acid
Synonyms: N-acetyl-D-tryptophan
Molecular Weight (g/mol): 246.27
Molecular Formula: C13H14N2O3
InChi Key: DZTHIGRZJZPRDV-GFCCVEGCSA-N
SMILES: CC(=O)N[C@H](CC1=CNC2=CC=CC=C12)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results