accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22830-45-1

IUPAC Name: λ²-iron(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) dihydrate
Synonyms: ferrous gluconate dihydrate
Molecular Weight (g/mol): 482.17
Molecular Formula: C12H26FeO16
InChi Key: OKGNXSFAYMSVNN-SYAJEJNSSA-L
SMILES: O.O.[Fe++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)