Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) Gluconate Hydrate 95.0+%, TCI America™
SDP

Catalog No. G0038500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G0038500G 500 g
G003825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G0038500G Supplier TCI America Supplier No. G0038500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22830-45-1
Molecular Formula C12H26FeO16
Molecular Weight (g/mol) 482.17
MDL Number MFCD00150872
InChI Key OKGNXSFAYMSVNN-SYAJEJNSSA-L
Synonym iron ii gluconate hydrate
PubChem CID 131851469
IUPAC Name λ²-iron(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) dihydrate
SMILES O.O.[Fe++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Formula Weight 446.14
Percent Purity ≥95.0% (T)
Chemical Name or Material Iron(II) Gluconate Hydrate
RTECSNumber LZ5180000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.