accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22831-39-6

IUPAC Name: dimagnesiosilane
Synonyms: magnesium silicide
Molecular Weight (g/mol): 76.7
Molecular Formula: Mg2Si
InChi Key: YTHCQFKNFVSQBC-UHFFFAOYSA-N
SMILES: [Mg]=[Si]=[Mg]
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)