Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium silicide, 99.99% (metals basis)

Catalog No. AA4551830
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4551830 250 g
AA4551809 10 g
AA4551818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4551830 Supplier Thermo Scientific Chemicals Supplier No. 04551830
Add to Cart

missing translation for 'validMainframeMsg'

Air and Moisture Sensitive; Decomposes in water, HCl | CAS: 22831-39-6 | Mg2Si | 76.695 g/mol

Magnesium silicide is used to produce alloys such as aluminum alloys. It is also used in food, beverages, as a flavor enhancer and as an intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22831-39-6
Molecular Formula Mg2Si
Molecular Weight (g/mol) 76.695
MDL Number MFCD00016202
InChI Key YTHCQFKNFVSQBC-UHFFFAOYSA-N
Synonym magnesium silicide, dimagnesium silicide, dimagnesium monosilicide, unii-475e6fmg3k, magnesiumsilicid, magnesium, compd. with silicon 2:1, magnesium silicide, powder, magnesium silicide-45 my, magnesium silicide un2624 dangerous when wet
PubChem CID 89858
SMILES [Mg]=[Si]=[Mg]

Specifications

Melting Point 1102°C
Color Blue
Physical Form Pieces
Quantity 250 g
Sensitivity Air and moisture sensitive
Assay Percent Range 99.99% (Metals basis)
Density 2
Solubility Information Insoluble in water and denser than water.
Formula Weight 76.71
Chemical Name or Material Magnesium silicide
Hazard Category H261
Precautionary Statement P223-P231+P232-P280-P335+P334-P370+P378q-P501c
DOTInformation DOT Class: 4.3, Packaging Group: II
EINECSNumber 245-254-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.