CAS RN 2284-96-0
IUPAC Name:
2-(4-fluorophenyl)-4,5-diphenyl-1H-imidazole
Synonyms:
2-(4-fluorophenyl)-4,5-diphenyl-1H-imidazole
Molecular Weight (g/mol):
314.36
Molecular Formula:
C21H15FN2
InChi Key:
NSXOBUOVKKFIHN-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-(4-Fluorophenyl)-4,5-diphenylimidazole 98.0+%, TCI America™
CAS: 2284-96-0 Molecular Formula: C21H15FN2 Molecular Weight (g/mol): 314.36 MDL Number: MFCD00389667 InChI Key: NSXOBUOVKKFIHN-UHFFFAOYSA-N PubChem CID: 800028 IUPAC Name: 2-(4-fluorophenyl)-4,5-diphenyl-1H-imidazole SMILES: FC1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=CC=C1)C1=CC=CC=C1