Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Fluorophenyl)-4,5-diphenylimidazole 98.0+%, TCI America™
SDP

Catalog No. F0968200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0968200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F0968200MG Supplier TCI America Supplier No. F0968200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2284-96-0
Molecular Formula C21H15FN2
Molecular Weight (g/mol) 314.36
MDL Number MFCD00389667
InChI Key NSXOBUOVKKFIHN-UHFFFAOYSA-N
PubChem CID 800028
IUPAC Name 2-(4-fluorophenyl)-4,5-diphenyl-1H-imidazole
SMILES FC1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 256°C
Color White-Yellow
Quantity 200 mg
Formula Weight 314.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Fluorophenyl)-4,5-diphenylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.