accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClONNClClCl

CAS RN 22916-47-8

IUPAC Name: 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole
Synonyms: miconazole monistat 7 combination pack daktarin talc aflorix andergin miconazole product daktarin (+-)-miconazole nitrate monistat IV (TN) miconazole nitrate preparation
Molecular Weight (g/mol): 416.12
Molecular Formula: C18H14Cl4N2O
InChi Key: BYBLEWFAAKGYCD-UHFFFAOYNA-N
SMILES: ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
Physical Form 
  • (4)
  • (2)
  • (4)
  • (2)
Percent Purity 
  • (4)
  • (3)
Grade 
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (1)
  • (3)
  • (4)
  • (5)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (4)
  • (4)
  • (2)