Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Miconazole, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18604590 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-590 1 kg Amber Glass Bottle
18-604-587 5 g Amber Glass Bottle
18-604-588 25 g Amber Glass Bottle
18-604-589 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-590 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M11881KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

M1188, 22916-47-8, C18H14Cl4N2O

Spectrum™ Chemical Miconazole, USP is a topical imidazole antifungal agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 22916-47-8
Molecular Formula C18H14Cl4N2O
Molecular Weight (g/mol) 416.12
MDL Number MFCD00216019
InChI Key BYBLEWFAAKGYCD-UHFFFAOYNA-N
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole
SMILES ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C(Cl)=C1

Specifications

CAS Max % 100%
Ignition Residue 0.2%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.