accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22921-58-0

IUPAC Name: 2-(propan-2-yloxy)benzaldehyde
Synonyms: 2-isopropoxybenzaldehyde
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: ZZJVNPRHHLLANO-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results