Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Isopropoxybenzaldehyde

Catalog No. CDS002654 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002654 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002654 Supplier Sigma Aldrich Supplier No. CDS00265450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-tert-butylcyclohexyl acetate has been evaluated as a fragrance ingredient and its toxicologic and dermatological analysis has been reported
  • Cis and trans forms of 4-tert-butylcyclohexyl acetate are widely used as a perfume for cosmetics including soaps.4-tert-butylcyclohexyl acetate may be used in the preparation of high loading fragrance encapsulation based on polymer-blend of ethylcellulose, hydroxypropyl methylcellulose and poly(vinyl alcohol)

Specifications

CAS 22921-58-0
Molecular Formula C10H12O2
Linear Formula C10H12O2
Quantity 50 mg
MDL Number MFCD01571834
Molecular Weight (g/mol) 164.2
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.