accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22921-68-2

IUPAC Name: 2-bromo-5-methoxybenzoate
Synonyms: 2-bromo-5-methoxybenzoate
Molecular Weight (g/mol): 230.04
Molecular Formula: C8H6BrO3
InChi Key: ODHJOROUCITYNF-UHFFFAOYSA-M
SMILES: COC1=CC=C(Br)C(=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)